Få mere at vide
Fistreem International™ Glass Water Still, Loughborough
Output flowhastighed:
4 l/time
8 l/time
Dimensioner (L x B x H):
450 x 390 x 820 mm
580 x 430 x 1040 mm
Elektriske krav:
220 V 50 Hz 3 kW
220 V 50 Hz 6 kW
240 V 50 Hz 3 kW
Pakningsstørrelse:
Hver
Højde (metrisk):
820 mm
1040 mm
Beskrivelse
This conventional style water still is available in two output sizes, 4L/hr and 8L/hr. Each produces a high quality pyrogen free distillate. The assembly and operation of these stills is simple and their well proven design ensures that the minimum of time is spent on their maintenance. The heavy duty chromium plated heaters are fitted with a thermostatic cut out which prevents damage to the glassware should the water supply be broken or fail completely. The cut out automatically resets when cool. A constant level device automatically maintains the boiler contents at the optimum level. A siphon tube fitted to the boiler enables easy descaling when necessary. With a bench/wall mounting stand.
Tekniske data
Tekniske data
| Type | Water Still, Loughborough |
| Output flowhastighed | 4 l/time |
| Dimensioner (L x B x H) | 450 x 390 x 820 mm |
| Elektriske krav | 240 V 50 Hz 3 kW |
| Spænding | 240 V |