Learn More
BRAND™ Glass Volumetric Flask
With glass stopper
Brand: BRAND™ 956978
Additional Details : Weight : 0.30000kg
Description
Specifications
Glass | |
Borosilicate Glass (3.3) | |
A |
50 mL | |
Ground Joint NS 12/21 |