Learn More
BRAND™ Glass Volumetric Flask
With glass stopper
Brand:  BRAND™ 956978
Additional Details : Weight : 0.30000kg
Product Code. 15941841
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Boro 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • With USP batch certificate
  • Specifications
    Specifications
    Glass
    Borosilicate Glass (3.3)
    A
    50 mL
    Ground Joint NS 12/21