Learn More
Hecht Karl™ Glass Vials
6.20 DKK - 1250.80 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10053941
|
Hecht Karl™
42783032 |
60 mL |
6.20 DKK
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10319311
|
Hecht Karl™
42783004 |
2 mL |
1250.80 DKK
Pack of 690 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Specifications
Glass | |
Plastic snap cap | |
Vial |