Learn More
Poulten Selfe™ Glass Ubbelohde
Suitable for transparent liquids
Brand: Poulten Selfe™ 1634/13
Additional Details : Weight : 1.00000kg
Description
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.Specifications
Ubbelohde Viscometer | |
Yes | |
4 |
2000 to 10,000 cSt | |
Glass |