Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand: Hirschmann™ 8900365
18.43 DKK valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
- Acc. to DIN 12 252
- Available to fit a range of sizes
Specifications
Stopper | |
Glass | |
NS 12/21 |
1/Ea. | |
For tight sealing of standard ground vessels |