Learn More
Duran™ Glass Stoppers
Manufactured from Borosilicate glass. DURAN™ Glass Stoppers are semi-hollow.
Brand:  Duran™ 286114804
1235.00 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 15901521
  • 1300.00 DKK / Pack of 100
In Stock
Explore more special offers
Description
  • DURAN™ cap
  • Flat head
  • Semi-hollow
  • Made of Borosilicate glass