Learn More
Trajan Scientific and Medical™ Glass Lined Tubing
Glass Lined Tubing (GLT) is borosilicate glass lined stainless steel tubing giving excellent resistance to the harshest of conditions
Brand:  Trajan Scientific and Medical™ 082718
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13140753
  • 555.00 DKK / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Excellent reistance to strong acids and bases
  • Ideal for HPLC columns - particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
Specifications
Tubing
0.5 mm
60 cm
500°C
1/16 in.
1.59 mm
Stainless Steel, Borosilicate Glass (Lining)