Learn More
Cytiva Glass Fiber GF10 Filter
For use as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units
Brand: Cytiva 10370393
669.75 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.17000kg
Description
This filter, with extreme mechanical stability and temperature resistance up to 180°C, is used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units.Specifications
12 s/100 mL/1.56 cm2 | |
Roll | |
Medium | |
Grade GF 10 | |
GF10 Filter |
Whatman Grade GF 10 glass microfiber filters from Cytiva's business are made from borosilicate glass and strengthened with an organic binder to provide high mechanical stability. | |
Borosilicate Glass | |
Used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units. | |
1 Each |