Få mere at vide
Glafenine, MedChemExpress
Beskrivelse
Glafenine is a non-steroidal anti-inflammatory drug (NSAID), is a non-narcotic analgesic agent, widely used for the treatment of pains of various origins.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Glafenine |
| CAS | 3820-67-5 |
| Farve | Light Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H17ClN2O4 |
| Mængde | 100 mg |
| Synonym | Glafenin |
| Opløselighedsinformation | DMSO : 100 mg/mL (268.24 mM; Need ultrasonic) |
| SMIL | O=C(OCC(O)CO)C1=CC=CC=C1NC2=CC=NC3=CC(Cl)=CC=C23 |
| Molekylvægt (g/mol) | 372.8 |
| Vis mere |