missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Glafenine hydrochloride, MedChemExpress
Description
Glafenine hydrochloride is a non-narcotic analgesic and non-steroidal anti-inflammatory drug. It is an ABCG2 inhibitor with an IC50 of 3.2 μM.
Specifications
Specifications
| Kemisk navn eller materiale | Glafenine hydrochloride |
| CAS | 65513-72-6 |
| Farve | Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H18Cl2N2O4 |
| Mængde | 100 mg |
| Synonym | Glafenin hydrochloride |
| Opløselighedsinformation | DMSO : 41.67 mg/mL (101.82 mM; Need ultrasonic) |
| SMIL | [H]Cl.O=C(OCC(O)CO)C1=CC=CC=C1NC2=CC=NC3=CC(Cl)=CC=C23 |
| Molekylvægt (g/mol) | 409.26 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?