missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GJC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-87499
This item is not returnable.
View return policy
Description
GJC3 Polyclonal specifically detects GJC3 in Human samples. It is validated for Western Blot.
Specifications
GJC3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
349149 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
connexin 29, connexin-30.2, Connexin-31.3, CX29, Cx30.2, CX30.2connexin-31.3, CX31.3, Gap junction epsilon-1 protein, gap junction gamma-3 protein, gap junction protein, epsilon 1, 29kDa, gap junction protein, gamma 3, 30.2kDa, GJE1 | |
The immunogen is a synthetic peptide directed towards the middle region of human GJC3 | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GJC3 Antibody, Novus Biologicals™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction