Få mere at vide
Tocris Bioscience™ GIT 27
GREENER_CHOICE
Artikelnummer. 18084652
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18084652 10 mg
18045422 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18084652 Leverandør Tocris Bioscience™ Leverandørnr. 3270/10
Immunomodulator; reduces production of pro-inflammatory cytokines
Orally active immunomodulatory agent that primarily targets macrophages. Inhibits TNF-α secretion via interference of macrophage toll-like receptor (TLR) 4 and TLR 2/6 signaling pathway. Also reduces the secretion of pro-inflammatory cytokines IL1-β, IL-10 and IFN-γ. Antidiabetogenic; prevents IL-β and IFN-γ-induced pancreatic islet cell death in vitro.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6501-72-0
Molekylær formel C11H11NO3
Tekniske data
Kemisk navn eller materiale GIT 27
Mængde 10 mg
Formel vægt 205.21
Renhed >99%
Molekylvægt (g/mol) 205.21