Få mere at vide
Ginsenoside RK2, TRC
Artikelnummer. 30240736
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240736 2.5 mg 2.5mg
30240593 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240736 Leverandør Toronto Research Chemicals Leverandørnr. TRCG4102302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 364779-14-6
Molekylær formel C36H60O7
Molekylvægt (g/mol) 604.86
Synonym (3β,12β)-12-Hydroxydammara-20,24-dien-3-yl β-D-glucopyranoside, 3β,12β-Dihydroxydammar-20(21),24-diene-3-O-β-D-glucopyranoside, PAN 20;
SMIL CC(=CCCC(=C)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(C)(C)[C@@H]4CC[C@@]23C)C
Tekniske data
Kemisk navn eller materiale Ginsenoside RK2
Mængde 2.5 mg