Få mere at vide
Ginsenoside Rg3, TRC
Artikelnummer. 30240898
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240898 25 mg
30240754 100 mg
30240886 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240898 Leverandør Toronto Research Chemicals Leverandørnr. TRCG38759025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14197-60-5
Molekylær formel C42 H72 O13
Molekylvægt (g/mol) 785.01
Synonym β-D-Glucopyranoside, (3β,12β)-12,20-dihydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-, Dammar-24-ene-12β,20-diol, 3β-[(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]- (8CI), Dammarane, β-D-glucopyranoside deriv., (20S)-Propanaxadiol, (3β,12β)-12,20-Dihydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, (3β,12β)-12,20-Dihydroxydammar-24-en-3-yl O-β-D-glucopyranosyl-(1→2)-β-D-glucopyranoside, 20(S)-Ginsenoside-Rg3, 20(S)-Rg3, 20S-Ginsenoside Rg3, 3-O-β-D-Glucopyranosyl-(1→2)-β-D-glucopyranosyldammar-24-ene-3β,12β,20S-triol, Ginsenoside 20-Rg3, Ginsenoside Rg3, S-Ginsenoside Rg3
IUPAC navn (2S,3R,4S,5S,6R)-2-[(2R,3R,4S,5S,6R)-4,5-dihydroxy-2-[[(3S,5R,8R,9R,10R,12R,13R,14R,17S)-12-hydroxy-17-[(2S)-2-hydroxy-6-methylhept-5-en-2-yl]-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-6-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL CC(=CCC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(C)(C)[C@@H]4CC[C@@]23C)C
Tekniske data
Kemisk navn eller materiale S-Ginsenoside Rg3
Mængde 25 mg
InChI formel InChI=1S/C42H72O13/c1-21(2)10-9-14-42(8,51)22-11-16-41(7)29(22)23(45)18-27-39(5)15-13-28(38(3,4)26(39)12-17-40(27,41)6)54-37-35(33(49)31(47)25(20-44)53-37)55-36-34(50)32(48)30(46)24(19-43)52-36/h10,22-37,43-51H,9,11-20H2,1-8H3/t22-,23+,24+,25+,26-,27+,28-,29-,30+,31+,32-,33-,34+,35+,36-,37-,39-,40+,41+,42-/m0/s1
Formel vægt 784.4973