missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Specifications
Specifications
| Kemisk navn eller materiale | Ginsenosid Rg1 |
| CAS | 22427-39-0 |
| Mængde | 100 mg |
| Smeltepunkt | 188 °C til 191 °C |
| Molekylær formel | C42 H72 O14 |
| Formel vægt | 801.02 |
| MDL nummer | MFCD00210293 |
| Fysisk form | Krystallinsk pulver eller krystaller |
| Farve | Farveløs eller hvid |
| Renhed | ≥90 % |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?