Få mere at vide
Gilson™ PIPETMAN Neo™ Multichannel Pipets
Shop alle Gilson produkter
Volumen (metrisk):
2 til 20 μl
20 til 200 μl
Antal kanaler:
8
12
Model-:
P12x200N
P12x20N
P8x200N
P8x20N
Pakningsstørrelse:
Hver
Beskrivelse
- Easy-to-remove tip ejector with autoclavable lower part
- Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
- PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
- Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
- Unique tip holder allows for a secure fit of most brands of tips
- Available in 8- and 12-channel models
Ordreinfo
Trade-in program available. Please contact your Fisher Scientific Sales representative for more details.
Tekniske data
Tekniske data
| Autoklaverbar | Autoclavable |
| Model- | P12x20N |
| Antal kanaler | 12 |
| Produktlinje | Pipetman Neo |
| Type | Multichannel Pipette |
| Volumen (metrisk) | 2 til 20 μl |
| garanti | 1 Year |