Få mere at vide
Gilson™ PIPETMAN™ M Pipettes
Shop alle Gilson produkter
Antal kanaler:
1
8
12
Model-:
P10mLM
P1200M
P12X10M
P12x100M
P12x1200M
P12x200M
P12x20M
P12x300M
P300M
P5000M
P8X20M
P8x100M
P8x10M
P8x1200M
P8x200M
P8x300M
Volumen (metrisk):
0,5 til 10 μl
1 til 10 ml
1 til 20 μl
10 til 10 μl
10 til 100 μl
10 til 300 μl
100 til 1200 μl
20 til 200 μl
20 til 300 μl
50 til 1200 μl
500 til 5000 μl
Kompatible tips:
D10 mL
D10, DL10, DF10 or DFL10 tips
D10, DL10, DF10, DFL10 tips
D1000, DF1000, D1200, DF1200
D1200
D200
D200, D300, DF100ST, DF200ST, DF300ST tips
D200, DF200, D300, DF300
D5000
DF100
DF1200
DL10, DF30ST, D200 tips
Pakningsstørrelse:
Hver
Beskrivelse
- Motorized piston with adjustable piston speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
- Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Specifications
| Kompatible tips | D200, D300, DF100ST, DF200ST, DF300ST tips |
| Farve | Yellow |
| Model- | P8x200M |
| Antal kanaler | 8 |
| Produktlinje | Pipetman |
| Type | Pipette |
| Volumen (metrisk) | 20 til 200 μl |
| Finger stil | Ergonomic |
| garanti | 1 Year |