missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-7 Recombinant Protein

Recombinant IL7 is a bioactive protein intended for use in cell culture applications

POA

Specifications

Purification Method Sequential Chromatography
Gene Alias IL-7
View More Specs

Description

Description

IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.

Approved for shipment on Wet or Dry Ice.

Specifications

Specifications

Sequential Chromatography
IL-7
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.