missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibberellin A9 Methyl Ester, TRC

Artikelnummer. 30240596
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240596 100 mg
30240729 50 mg
30240662 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Artikelnummer. 30240596 Leverandør Toronto Research Chemicals Leverandørnr. TRCG377325100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2112-08-5
Molekylær formel C20H26O4
Molekylvægt (g/mol) 330.42
Synonym GA9 Methyl Ester, 1,4a-Lactone-4a-hydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibbane-1alpha,10beta-dicarboxylic Acid Methyl Ester;
SMIL COC(=O)[C@H]1[C@@H]2[C@@]3(C)CCC[C@@]2(OC3=O)[C@@H]4CC[C@@H]5C[C@]14CC5=C

Tekniske data

Kemisk navn eller materiale Gibberellin A9 Methyl Ester
Mængde 100 mg
InChI formel InChI=1S/C20H26O4/c1-11-9-19-10-12(11)5-6-13(19)20-8-4-7-18(2,17(22)24-20)15(20)14(19)16(21)23-3/h12-15H,1,4-10H2,2-3H3/t12-,13-,14-,15-,18-,19+,20-/m1/s1
Formel vægt 330.1831
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.