Få mere at vide
Gibberellin A7, TRC
Artikelnummer. 30011332
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011332 50 mg
30006822 5 mg
30003529 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011332 Leverandør Toronto Research Chemicals Leverandørnr. TRCG37746050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 510-75-8
Molekylær formel C19 H22 O5
Molekylvægt (g/mol) 330.38
Synonym 1H-7,9a-Methanobenz[a]azulene-1,10-dicarboxylic acid, 2,4a,4b,5,6,7,8,9,10,10a-decahydro-2,4a-dihydroxy-1-methyl-8-methylene-, 1,4a-lactone (7CI), 4aα,4bβ-Gibb-3-ene-1α,10β-dicarboxylic acid, 2β,4a-dihydroxy-1-methyl-8-methylene-, 1,4a-lactone (8CI), Gibberellin A7 (6CI), 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibb-3-ene-1,10-dicarboxylic acid deriv., GA7, Gibberellin A7
SMIL C[C@]12[C@@H](O)C=CC3(OC1=O)[C@@H]4CC[C@@H]5CC4(CC5=C)[C@H]([C@H]23)C(=O)O
Tekniske data
Kemisk navn eller materiale Gibberellin A7
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C19H22O5/c1-9-7-18-8-10(9)3-4-11(18)19-6-5-12(20)17(2,16(23)24-19)14(19)13(18)15(21)22/h5-6,10-14,20H,1,3-4,7-8H2,2H3,(H,21,22)/t10-,11-,12+,13-,14-,17-,18?,19?/m1/s1
Formel vægt 330.1467
Procent renhed >95%