Få mere at vide
Gibberellin A7 Methyl Ester, TRC
Artikelnummer. 30240788
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240788 10 mg
30240970 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Artikelnummer. 30240788 Leverandør Toronto Research Chemicals Leverandørnr. TRCG37748010MG
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5508-47-4
Molekylær formel C20H24O5
Molekylvægt (g/mol) 344.4
Synonym 2beta,4a-Dihydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibb-3-ene-1alpha,10beta-dicarboxylic Acid 1,4a-Lactone Methyl Ester, Gibberellin A7 Methyl Ester, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene Gibb-3-ene-1,10-dicarboxylic Acid Deriv.;
SMIL COC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](O)C=C[C@@]2(OC3=O)[C@@H]4CC[C@@H]5C[C@]14CC5=C
Tekniske data
Kemisk navn eller materiale Gibberellin A7 Methyl Ester
Mængde 10 mg
InChI formel InChI=1S/C20H24O5/c1-10-8-19-9-11(10)4-5-12(19)20-7-6-13(21)18(2,17(23)25-20)15(20)14(19)16(22)24-3/h6-7,11-15,21H,1,4-5,8-9H2,2-3H3/t11-,12-,13+,14-,15-,18-,19+,20-/m1/s1
Formel vægt 344.1624