Få mere at vide
Gibberellic Acid Acetoxymethyl Ester, TRC
Artikelnummer. 30240514
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240514 2.5 g 2.5g
30240481 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Artikelnummer. 30240514 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3775002.5G
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1373154-68-7
Molekylær formel C22 H26 O8
Molekylvægt (g/mol) 418.44
Synonym Gibberellic Acid Acetoxymethyl Ester, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene-10-acetic acid, 1,2,4b,5,6,7,8,9,10,10a-decahydro-2,7-dihydroxy-1-methyl-8-methylene-13-oxo-, (acetyloxy)methyl ester, (1S,2S,4aR,4bR,7S,9aS,10S,10aR)-
SMIL CC(=O)OCOC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](O)C=C[C@@]2(OC3=O)[C@@H]4CC[C@]5(O)C[C@]14CC5=C
Tekniske data
Kemisk navn eller materiale Gibberellic Acid Acetoxymethyl Ester
Renhedsgrad noter HPLC
Mængde 2.5 g
InChI formel InChI=1S/C22H26O8/c1-11-8-20-9-21(11,27)6-4-13(20)22-7-5-14(24)19(3,18(26)30-22)16(22)15(20)17(25)29-10-28-12(2)23/h5,7,13-16,24,27H,1,4,6,8-10H2,2-3H3/t13-,14+,15-,16-,19-,20+,21+,22-/m1/s1
Formel vægt 418.1628
Procent renhed >95%