missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic acid, 90+%, Thermo Scientific Chemicals

Catalog Number p-7023863
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-QTWFBFKQSA-N
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
91757643
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
This item is not returnable. View return policy
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-QTWFBFKQSA-N
gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
91757643
(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O

It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a plant growth regulating hormone that promotes plant cell growth and elongation. They promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. They are also involved in processes such as gravitropism, tensioning and floral display. Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media.

Solubility
Soluble in water 20°C 5g/L. Soluble in Ethanol: 200 mg/4ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 222°C (decomposition)
Quantity 1 g
Beilstein 54346
Merck Index 14,4419
Solubility Information Soluble in water 20°C 5g/L. Soluble in Ethanol: 200mg/4ml.
Optical Rotation +78° (c=2 in Methanol)
Formula Weight 346.37
Percent Purity ≥90%
Chemical Name or Material Gibberellic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'