Få mere at vide
Gibberellic Acid-13C,d2 (90%), TRC
Artikelnummer. 30405716
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30405716 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405716 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3774531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Gibberellic Acid 13C D2 (17-13C 17,17-D2)
CAS 120396-36-3
Renhedsgrad noter HPLC
Molekylær formel 13C C18 D2 H20 O6
Mængde 1 mg
Synonym Gibberellic Acid-13C,d2, Gibberellic Acid 13C D2 (17-13C 17,17-D2)
InChI formel InChI=1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1/i1+1D2
SMIL [2H][13C](=C1C[C@]23C[C@@]1(O)CC[C@H]2[C@]45OC(=O)[C@](C)([C@@H](O)C=C4)[C@H]5[C@@H]3C(=O)O)[2H]
Molekylvægt (g/mol) 349.379
Formel vægt 349.158
Procent renhed >95