missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFPT1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99692AF750
Additional Details : Weight : 0.00970kg
Description
GFPT1 Polyclonal specifically detects GFPT1 in Human samples. It is validated for ELISA.Specifications
GFPT1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 750 | |
glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG | |
Produced in rabbits immunized with purified, recombinant GFPT1 (AAA58502.1; Gln 332-Glu 699). | |
0.1 mL | |
metabolism | |
2673 | |
Store at 4°C in the dark. | |
IgG |