Få mere at vide
Invitrogen™ GFP Monoclonal Antibody (GF28R), DyLight™ 680
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515256D680
Beskrivelse
MA5-15256-D680 has been successfully used in immunofluorescence and Western blot applications. DyLight 680 has an excitation/emission of 692/712 nm.
The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Tekniske data
GFP | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
GF28R | |
DyLight 680 | |
GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP | |
GFP from the jellyfish Aequorea Victoria N-terminal peptide-KLH conjugated. | |
50 μL | |
Primary | |
4°C, do not freeze | |
Liquid | |
Tag |
Certifikater
Der kræves et partinummer for at vise resultater for certifikater. For at finde dit varenummer på tidligere ordrer, skal du bruge vores ordrestatusområde.
Partinummer | Certifikattype | Dato | Katalognummer |
---|---|---|---|
3214127 | Analysecertifikat | 11/07/2025 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.