missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
GFP Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-308PE
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry, Knockdown Validated.
Tekniske data
GFP | |
Polyclonal | |
Flow Cytometry, KnockDown Validated | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. | |
0.1 mL | |
Primary | |
Non-species specific, C. elegans, Plant | |
IgG |
Flow Cytometry, KnockDown | |
PE | |
PBS with 0.05% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
No reaction was observed against Human, Mouse or Rat serum proteins. | |
Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
GFP Antibody, PE, Novus Biologicals™ > 0.1 mL; PE
Ser du en mulighed for forbedring?Del en indholdskorrektion