Få mere at vide
Gestodene-2,2,4,6,6,10-d6, CDN
Artikelnummer. 30310657
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310657 0.005 g
30308839 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310657 Leverandør CDN Leverandørnr. CDND65940.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Gestodene-2,2,4,6,6,10-D6
CAS 1542211-40-4
Molekylær formel C21 D6 H20 O2
Mængde 0.005 g
Synonym 18,19-Dinorpregna-4,15-dien-20-yn-3-one-2,2,4,6,6,10-d6, 13-ethyl-17-hydroxy-, (17α)- (ACI), (17α)-13-Ethyl-17-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one-2,2,4,6,6,10-d6 (ACI), Gestodene-D6, Gestodene-2,2,4,6,6,10-D6, Gestodene-d6, Gestodene-2,2,4,6,6,10-d6
InChI formel InChI=1 S/C21H26O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,10,12-13,16-19,23 H,3,5-9,11H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1/i5D2,6D2,13 D,16 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(CC)[C@H]3 C=C[C@@]4(O)C#C)[C@@]2([2 H])CC([2 H])([2 H])C1=O
IUPAC navn (8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6,10-hexadeuterio-13-ethyl-17-ethynyl-17-hydroxy-7,8,9,11,12,14-hexahydro-1 H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 316.4669
Formel vægt 316.2309 g/mol
Procent renhed 96 atom % D, min 98% Chemical Purity