missing translation for 'onlineSavingsMsg'
Learn More

Germanium solution for ICP 1g/L HNO3/HFmatrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical S022500

Additional Details : Weight : 0.50000kg Transport : UN number : 1760 Chem class : 8 Pack group : II


Packaging Bottle
Quantity 500mL

 View more versions of this product

Product Code. 12953414

  • 758.00 DKK / 500mL

Estimated Shipment
19-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

Specifications

1±0.002g/L
Ge
500mL
Bottle
HNO3/ HF
Mass spectrometry standards
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-element Standard Solution for ICP. Germanium (Ge) 1000mg/l in HNO3 5%,HF 1%

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P501-Dispose of contents/container to .

SDS
Documents