missing translation for 'onlineSavingsMsg'
Få mere at vide

Germanium(IV) Chloride, Sigma-Aldrich™

Artikelnummer. 15899090
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15899090 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Artikelnummer. 15899090 Leverandør Sigma Aldrich Leverandørnr. 8040790025
 Begrænset vare
Forventet forsendelse: 26-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Tekniske data

Kogepunkt 83.1°C
Kemisk navn eller materiale Germanium(IV) Chloride
Smeltepunkt -49.5°C
CAS 10038-98-9
Mængde 25 mL
Tæthed 1.88 g/cm3 at 20°C
MDL nummer MFCD00011029
Synonym Germanium(IV) chloride, Germanium tetrachloride, Germanium(IV) chloride
Molekylvægt (g/mol) 214.45
Grad Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.