Få mere at vide
Germacrene D (~90%) (Stabilized with hydroquinone), TRC
Artikelnummer. 17355353
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17355353 1 mg
30004452 25 mg
17935065 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17355353 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3677651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23986-74-5
Molekylær formel C15H24
Molekylvægt (g/mol) 204.35
Synonym [S-(E,E)]-1-Methyl-5-methylene-8-(1-methylethyl)-1,6-cyclodecadiene, (-)-Germacra-1(10),4(15),5-triene, (-)-Germacrene D, Germacrene D, [s-(E,E)]-1-Methyl-5-methylene-8-(1-methylethyl)-1,6-cyclodecyldidiene;
IUPAC navn (1E,6E,8S)-1-methyl-5-methylidene-8-propan-2-ylcyclodeca-1,6-diene
SMIL CC([C@H]1/C=C/C(CC/C=C(C)/CC1)=C)C
Tekniske data
Kemisk navn eller materiale Germacrene D
Mængde 1 mg
InChI formel InChI=1S/C15H24/c1-12(2)15-10-8-13(3)6-5-7-14(4)9-11-15/h7-8,10,12,15H,3,5-6,9,11H2,1-2,4H3/b10-8+,14-7+/t15-/m0/s1
Formel vægt 204.1878