missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranyl bromide, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.094 |
Boiling Point | 99°C to 101°C (10 mmHg) |
Flash Point | 95°C (203°F) |
Refractive Index | 1.504 |
Quantity | 5 g |
Beilstein | 1422412 |
Sensitivity | Light sensitive |
Solubility Information | Immiscible with water. |
Formula Weight | 217.16 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- Geranyl bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : 1-bromo-3,7-dimethyl-2,6-Octadiene
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'