Få mere at vide
Gentamicin X2 Sulfate, TRC
Artikelnummer. 30240761
Skift visning
Mængde:
0.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240761 5 mg
30240487 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240761 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3606055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57793-88-1
Molekylær formel C19H38N4O10.x(H2SO4)
Molekylvægt (g/mol) 580.6
Synonym O-2-Amino-2-deoxy-alpha-D-glucopyranosyl-(1-4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl-(1-6)]-2-deoxy-D-streptamine Sulfate
IUPAC navn (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMIL CCOc1ccc2C=CC(=O)Oc2c1
Tekniske data
Kemisk navn eller materiale Gentamicin X2 Sulfate
Mængde 5 mg
InChI formel InChI=1S/C19H38N4O10.H2O4S/c1-19(29)5-30-18(13(28)16(19)23-2)33-15-7(21)3-6(20)14(12(15)27)32-17-9(22)11(26)10(25)8(4-24)31-17;1-5(2,3)4/h6-18,23-29H,3-5,20-22H2,1-2H3;(H2,1,2,3,4)/t6-,7+,8+,9+,10+,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m0./s1
Formel vægt 580.2262