missing translation for 'onlineSavingsMsg'
Få mere at vide

Gentamicin C1 Deuterated Pentaacetate Salt (>90%), TRC

Artikelnummer. 30389645
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30389645 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30389645 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3605875MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Gentamicin C1 Deuterated Pentaacetate Salt
CAS 287730-69-2
Molekylær formel C31H63N5O17 (for non-Deuterated)
Mængde 5 mg
Synonym O-2-Amino-2,3,4,6,7-pentadeoxy-6-(methylamino)-α-D-ribo-heptopyranosyl-(14)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(16)]-2-deoxy-D-streptamine Pentaacetate Salt Deuterated
InChI formel InChI=1S/C21H43N5O7/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20/h9-20,25-29H,5-8,22-24H2,1-4H3/t9?,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+/m1/s1
SMIL CC(NC)[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O[C@@H]3[C@H](O)[C@@H](NC)[C@@](C)(O)CO3)[C@H](N)C[C@@H]2N)[C@H](N)CC1
Molekylvægt (g/mol) 777.86
Formel vægt 477.3162
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.