Få mere at vide
Genistein-d4 (4-hydroxyphenyl-2,3,5,6-d4), CDN
Artikelnummer. 30322988
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322988 0.01 g
30312599 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322988 Leverandør CDN Leverandørnr. CDND62820.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Genistein-2',3',5',6'-d4
CAS 187960-08-3
Molekylær formel C15 2H4 H6 O5
Mængde 0.01 g
InChI formel InChI=1 S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C2=COc3cc(O)cc(O)c3C2=O)c([2 H])c([2 H])c1O
IUPAC navn 5,7-dihydroxy-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)chromen-4-one
Molekylvægt (g/mol) 274.26
Formel vægt 274.0779 g/mol
Procent renhed 95 atom % D, min 97% Chemical Purity