Få mere at vide
Genistein-2’,3’,5’,6’-d4 (Major), TRC
Artikelnummer. 30240897
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240897 1 mg
30240551 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240897 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3500101MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 187960-08-3
Molekylær formel C15 2H4 H6 O5
Molekylvægt (g/mol) 274.26
Synonym 4H-1-Benzopyran-4-one, 5,7-dihydroxy-3-(4-hydroxyphenyl-2,3,5,6-d4)- (9CI, ACI), 5,7-Dihydroxy-3-(4-hydroxyphenyl-2,3,5,6-d4)-4H-1-benzopyran-4-one (ACI), Genistein-2',3',5',6'-d4, Genistein-d4 (4-hydroxyphenyl-2,3,5,6-d4), Genistein-2',3',5',6'-D4, 5,7-Dihydroxy-3-(4-hydroxyphenyl-d4)-4H-1-benzopyran-4-one
IUPAC navn 5,7-dihydroxy-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)chromen-4-one
SMIL [2H]c1c([2H])c(C2=COc3cc(O)cc(O)c3C2=O)c([2H])c([2H])c1O
Tekniske data
Kemisk navn eller materiale Genistein-2',3',5',6'-d4
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H/i1D,2D,3D,4D
Formel vægt 274.0779
Procent renhed >95%