missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ GeneArt™ Platinum™ Cas9 Nuclease (3 μg/μL)

Artikelnummer. 15425733
Skift visning
Klik for at se tilgængelige muligheder
:
75 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15425733 75 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
For research use only. Not for use in diagnostic procedures.
Artikelnummer. 15425733 Leverandør Invitrogen™ Leverandørnr. B25641

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy
For research use only. Not for use in diagnostic procedures.

Indeholder

1 vial GeneArt Platinum Cas9 Protein

Maximum efficient, the Cas9 RNP complex minimizes off-target cleavage compared to vector-based systems.

Maximum efficient, the Cas9 RNP complex minimizes off-target cleavage compared to vector-based systems.

GeneArt Platinum Cas9 Nuclease is the recombinant Streptococcus pyogenes Cas9 (wt) protein purified from E. coli that can be used for genome editing with CRISPR technology. Cas9 protein forms a very stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. The Cas9 RNP complex can act immediately once it enters the cell since there are no requirements for transcription or translation as with vector- or mRNA-based systems. Additionally, the Cas9 RNP complex is rapidly cleared from the cell minimizing the chance of off-target cleavage when compared to other systems (Kim, et al. 2014). Incorporation of a nuclear localization signal (NLS) aids delivery to the nucleus, thus increasing the rate of genomic DNA cleavage.

GeneArt Platinum Cas9 Nuclease features include:
  • Streamline cell engineering by eliminating transcription and translation in the cell
  • Eliminate time consuming cloning steps
  • Minimize off-target cleavage due to rapid clearance of the protein complex from the cell

Certificeringer

CoA

Overholdelse

CoA

Anbefalet opbevaring

Store at -5 to -30°C

TRUSTED_SUSTAINABILITY

Tekniske data

Koncentration 3 μg/μL
Indhold og opbevaring 1 vial of GeneArt platinum Cas9 protein. Store at -5 to -30°C
Oversigt GeneArt™ Platinum™ Cas9 Nuclease, 75μg, 3μg/μL Concentration, –5 to –30°C Storage Temperature, DNA Replication and Repair Research Category, For Research Use Only, Dry Ice Shipping Condition
Produkttype GeneArt™ Platinum™ Cas9 nuclease
Mængde 75 μL, 75 μg
Til brug med (applikation) Genetic Disease Research
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.