Få mere at vide
Gemfibrozil Impurity E (as (E)-Isomer), Mikromol™
Artikelnummer. 30459740
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459740 4 x 25 mg
30052456 100 mg
30050214 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459740 Leverandør Mikromol Leverandørnr. MM0641.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Gemfibrozil Impurity E (as (E)-Isomer), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-[2,5-Dimethyl-4-[(E)-prop-1-enyl]phenoxy]-2,2-dimethylpentanoic Acid
CAS 1315604-10-4
Analyt- eller komponentnavne 5-[2,5-Dimethyl-4-[(E)-prop-1-enyl]phenoxy]-2,2-dimethylpentanoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C18 H26 O3
Certificeringer/compliance ISO 17025
Synonym 5-[2,5-Dimethyl-4-(prop-1-enyl)phenoxy]-2,2-dimethylpentanoic Acid, Gemfibrozil Imp. E (EP), 5-[2,5-Dimethyl-4-(prop-1-enyl)phenoxy]-2,2-dimethylpentanoic Acid, 5-[2,5-Dimethyl-4-[(E)-prop-1-enyl]phenoxy]-2,2-dimethylpentanoic Acid, Gemfibrozil Impurity E as (E)-Isomer
InChI formel InChI=1S/C18H26O3/c1-6-8-15-11-14(3)16(12-13(15)2)21-10-7-9-18(4,5)17(19)20/h6,8,11-12H,7,9-10H2,1-5H3,(H,19,20)/b8-6+
Løsningstype Neat
SMIL C\C=C\c1cc(C)c(OCCCC(C)(C)C(=O)O)cc1C
IUPAC navn 5-[2,5-dimethyl-4-[(E)-prop-1-enyl]phenoxy]-2,2-dimethyl-pentanoic acid
Molekylvægt (g/mol) 290.4
Mængde 4 x 25 mg
Formel vægt 290.1882 g/mol