Få mere at vide
Gemcitabine Monophosphate, TRC
Artikelnummer. 30009064
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009064 1 mg
30009013 5 mg
30007453 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009064 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3050051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 116371-67-6
Molekylær formel C9 H12 F2 N3 O7 P
Molekylvægt (g/mol) 343.18
Synonym 5'-Cytidylic acid, 2'-deoxy-2',2'-difluoro-, 2'-Deoxy-2',2'-difluoro-5'-cytidylic acid, 2',2'-difluorodeoxycytidine monophosphate, Gemcitabine 5'-phosphate, Gemcitabine monophosphate
IUPAC navn [(2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-4,4-difluoro-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMIL NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)(O)O)[C@@H](O)C2(F)F
Tekniske data
Kemisk navn eller materiale Gemcitabine Monophosphate
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C9H12F2N3O7P/c10-9(11)6(15)4(3-20-22(17,18)19)21-7(9)14-2-1-5(12)13-8(14)16/h1-2,4,6-7,15H,3H2,(H2,12,13,16)(H2,17,18,19)/t4-,6-,7-/m1/s1
Formel vægt 343.0381
Procent renhed >95%