Få mere at vide
Biodesign™ Gel Protector Sheet Gelfile
Beskrivelse
Stores, organises and displays dried polyacrylamide and agarose gels
- Each BioDesign GelFile™ binder contains heavy crystal clear vinyl pages with pockets and white backgrounds
- The pages are chemically neutral and are guaranteed not to alter your dried gels or turn yellow for TEN years
- Dried gels are held flat for long term storage without curling
- Shields gels from moisture, accidental spills and possible damage
- White background allows optimal comparison between gels
- Each BioDesign GelFile™ binder holds 20 to 25 pages
- A clear label holder on the spine allows you to personalise each binder
Tekniske data
Tekniske data
| Gel størrelse | 11,5 x 18 cm |
| Mængde | 20 Pages (Stores 80 Gels) |
| Produkttype | Gel Protector Sheets |