Learn More
Fisherbrand™ Gel Casting Unit
Allows three gels to be cast simultaneously
Brand: Fisherbrand™ HU13-CU
This item is not returnable.
View return policy
Description
- Silicone gaskets to prevent leaks
- Seated in groove of each casting tray form leakfree seal against the walls of casting unit
- Allows three gels to be cast simultaneously
Specifications
Rapid Electrophoresis | |
1 Each | |
12.5 cm |
Gel Casting Unit | |
15 cm |
Product Content Correction
Product Title
Fisherbrand™ Gel Casting Unit > Gel Casting Unit
Spot an opportunity for improvement?Share a Content Correction