Få mere at vide
Gefarnate (Mixture of Isomers), TRC
Artikelnummer. 30135003
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135003 500 mg -
30132620 2.5 g 2.5g
30008085 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135003 Leverandør Toronto Research Chemicals Leverandørnr. TRCG303500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51-77-4
Molekylær formel C27 H44 O2
Molekylvægt (g/mol) 400.64
Synonym 4,8,12-Tetradecatrienoic acid, 5,9,13-trimethyl-, (2E)-3,7-dimethyl-2,6-octadien-1-yl ester, (4E,8E)-, 4,8,12-Tetradecatrienoic acid, 5,9,13-trimethyl-, (2E)-3,7-dimethyl-2,6-octadienyl ester, (4E,8E)- (9CI), 4,8,12-Tetradecatrienoic acid, 5,9,13-trimethyl-, 3,7-dimethyl-2,6-octadienyl ester, (E,E,E)- (8CI), 5,9,13-Trimethyl-4,8,12-tetradecatrienoic acid 3,7-dimethyl-2,6-octadienyl ester, Alsanate, Arsanyl, DA 688, Dixnalate, Gefanil, Gefarnate, Gefarnil, Gefarnyl, Gefulcer, Geranyl farnesylacetate, Osteol, Salanil, Zackal, trans-3,7-Dimethyl-2,6-octadienyl 5,9,13-trimethyl-4,8,12-tetradecatrienoate
IUPAC navn [(2E)-3,7-dimethylocta-2,6-dienyl] (4E,8E)-5,9,13-trimethyltetradeca-4,8,12-trienoate
SMIL CC(=CCC\C(=C\CC\C(=C\CCC(=O)OC\C=C(/C)\CCC=C(C)C)\C)\C)C
Tekniske data
Kemisk navn eller materiale Gefarnate
Mængde 500 mg
InChI formel InChI=1S/C27H44O2/c1-22(2)12-8-14-24(5)16-10-17-25(6)18-11-19-27(28)29-21-20-26(7)15-9-13-23(3)4/h12-13,16,18,20H,8-11,14-15,17,19,21H2,1-7H3/b24-16+,25-18+,26-20+
Formel vægt 400.3341