Learn More
Gefapixant, MedChemExpress
Product Code. 16493438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493438 10 mg 10mg
16492766 25 mg 25mg
16446167 5 mg 5mg
16414656 100 mg 100mg
16409956 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493438 Supplier Medchem Express Supplier No. HY101588
 Restricted Item
Forventet forsendelse: 30-11-2026
MedChemExpress Gefapixant is an orally active and potent purinergic P2X3 receptor (P2X3R) antagonist, with IC50 values of ∼30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Gefapixant can be used for the research of chronic cough and knee osteoarthritis.
Gefapixant is an orally active and potent purinergic P2X3 receptor (P2X3R) antagonist, with IC50 values of ∼30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Gefapixant can be used for the research of chronic cough and knee osteoarthritis.
Specifications
Chemical Name or Material Gefapixant
CAS 1015787-98-0
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O4S
Quantity 10 mg
Synonym MK-7264 AF-219
Solubility Information DMSO : 5 mg/mL (14.15 mM; ultrasonic and adjust pH to 5-6 with HCl)
SMILES O=S(C1=CC(OC2=CN=C(N)N=C2N)=C(C(C)C)C=C1OC)(N)=O
Molecular Weight (g/mol) 353.4
Formula Weight 353.4
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus