Learn More
Cytiva Spacers
Brand: Cytiva SE2619T-2-.75
Additional Details : Weight : 0.50000kg
Specifications
Gray | |
Spacer | |
2/Pk. | |
0.02 in. |
SE 250, 260 and MiniVE | |
PVC | |
T-shaped | |
0.75 mm |