missing translation for 'onlineSavingsMsg'
Få mere at vide

GDP366, MedChemExpress

Artikelnummer. 16492418
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16492418 5 mg 5mg
16470219 10 mg 10mg
16435437 50 mg -
16419306 100 mg -
16414086 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16492418 Leverandør Medchem Express Leverandørnr. HYU00177

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress GDP366, a dual inhibitor of survivin and Op18, induces cell growth inhibition, cellular senescence and mitotic catastrophe in human cancer cells.

GDP366, a dual inhibitor of survivin and Op18, induces cell growth inhibition, cellular senescence and mitotic catastrophe in human cancer cells.

Tekniske data

Kemisk navn eller materiale GDP366
CAS 501698-03-9
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C20H17N5OS
Mængde 5 mg
Opløselighedsinformation DMSO : 62.5 mg/mL (166.47 mM; Need ultrasonic)
SMIL O=C(NC1=CC=CC(C)=C1)NC2=CC=C(C3=CSC4=NC=NC(N)=C43)C=C2
Molekylvægt (g/mol) 375.45
Formel vægt 375.45
Procent renhed 99.73%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.