missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies GDC-0941

Artikelnummer. 17120182
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17120182 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17120182 Leverandør STEMCELL Technologies Leverandørnr. 73152

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

PI3K/AKT pathway inhibitor; Inhibits Class I PI3Ks

  • GDC-0941 is an inhibitor of phosphatidylinositol-4,5-bisphosphate 3-kinase (PI3K)
  • It inhibits class I catalytic subunits of PI3K p110α, β, δ, and γ with IC50 values of 3, 33, 3, and 75 nM, respectively, by binding near the ATP binding pocket (Folkes et al.; Berndt et al.) It shows selectivity against class II, III, and IV PI3K isoforms as well, but with lower efficacy; for example, it inhibits phosphatidylinositol-4-phosphate 3-kinase C2 domain subunit β (C2β, Class II) and mammalian target of rapamycin (mTOR, Class IV) in the high nanomolar range and most others in the micromolar range (Folkes et al.)
  • Inhibits proliferation of multiple cancer cell lines, such as U87MG (glioblastoma), PC3 (prostate) and MDA-MB-361 (breast) lines, in vitro and in mouse xenograft models (Folkes et al.; Raynaud et al.; O'Brien et al.)
  • Induces apoptosis and inhibits xenograft tumor growth in combination with a MAP/ERK kinase (MEK) inhibitor GDC-0973 (Hoeflich et al.)

Tekniske data

CAS 957054-30-7
Synonym GNE 0941; Pictilisib; Pictrelisib
Mængde 10 mg
Molekylær formel C23H27N7O3S2
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.