Få mere at vide
GDC-0575 dihydrochloride, MedChemExpress
Artikelnummer. 16494588
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494588 10 mg 10mg
16485436 100 mg -
16480887 50 mg -
16453146 25 mg -
16447307 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16494588 Leverandør Medchem Express Leverandørnr. HY112167A
MedChemExpress GDC-0575 dihydrochloride (ARRY-575 dihydrochloride) is an orally bioavailable CHK1 inhibitor, with an IC50 of 1.2 nM, and has antitumor activity.
GDC-0575 dihydrochloride (ARRY-575 dihydrochloride) is an orally bioavailable CHK1 inhibitor, with an IC50 of 1.2 nM, and has antitumor activity.
Tekniske data
Kemisk navn eller materiale GDC-0575 dihydrochloride
CAS 1657014-42-0
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C16H22BrCl2N5O
Mængde 10 mg
Synonym ARRY-575 dihydrochloride RG7741 dihydrochloride
Opløselighedsinformation DMSO : 65 mg/mL (144.06 mM; Need ultrasonic) ∣H2O : 25 mg/mL (55.41 mM; Need ultrasonic)
SMIL O=C(C1CC1)NC2=CNC3=NC=C(Br)C(N4C[C@H](N)CCC4)=C32.[2HCl]
Molekylvægt (g/mol) 451.19
Formel vægt 451.19
Procent renhed 99.32%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease