missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ GCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523706
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody is predicted to react with rat based on sequence homology.
The protein encoded by this gene is a peripheral membrane protein localized to the trans-Golgi network. It is sensitive to brefeldin A. This encoded protein contains a GRIP domain which is thought to be used in targeting. Alternative splicing results in multiple transcript variants.
Tekniske data
| GCC2 | |
| Polyclonal | |
| Unconjugated | |
| GCC2 | |
| 0610043A03Rik; 185 kDa Golgi coiled-coil protein; 2210420P05Rik; 2600014C01Rik; AW112121; CLL-associated antigen KW-11; CTCL tumor antigen se1-1; GCC protein, 185-kD; GCC185; GCC2; Golgi coiled-coil protein GCC185; GRIP and coiled-coil domain containing 2; GRIP and coiled-coil domain-containing protein 2; KIAA0336; mKIAA0336; Ran-binding protein 2-like 4; RanBP2L4; REN53; Renal carcinoma antigen NY-REN-53 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 70297, 9648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.50 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8CHG3, Q8IWJ2 | |
| GCC2 | |
| KLH conjugated synthetic peptide between 1581-1609 amino acids from the C-terminal region of human GCC2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion