missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ GCC2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142284
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Additional Information: This antibody is expected to recognise both reported isoforms (NP_055450.1and NP_852118.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
The protein encoded by this gene is a peripheral membrane protein localized to the trans-Golgi network. It is sensitive to brefeldin A. This encoded protein contains a GRIP domain which is thought to be used in targeting. Alternative splicing results in multiple transcript variants.
Tekniske data
| GCC2 | |
| Polyclonal | |
| Unconjugated | |
| GCC2 | |
| 0610043A03Rik; 185 kDa Golgi coiled-coil protein; 2210420P05Rik; 2600014C01Rik; AW112121; CLL-associated antigen KW-11; CTCL tumor antigen se1-1; GCC protein, 185-kD; GCC185; GCC2; Golgi coiled-coil protein GCC185; GRIP and coiled-coil domain containing 2; GRIP and coiled-coil domain-containing protein 2; KIAA0336; mKIAA0336; Ran-binding protein 2-like 4; RanBP2L4; REN53; Renal carcinoma antigen NY-REN-53 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 9648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IWJ2 | |
| GCC2 | |
| Peptide with sequence C-QKLTNKNNKIED. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion