Learn More
Trajan Scientific™ Shimadzu Plug Style Premium High-Temperature HT Septa
Retains softness and pierceability at high temperatures and low injection port adhesion.
Brand:  Trajan Scientific™ 041895
Additional Details : Weight : 0.50000kg
Product Code. 12995721
  • 1275.00 DKK / Pack of 50
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Description
Description
  • Bleed and temperature optimized
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
Specifications
BTO Silicone
400°C
HT GC Plug
50/Pk.